Koudespray
Leverancier: HM Industrieservice
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
291-1203EA 41.6 EUR
291-1203
Koudespray
Vriessprays
For applications in the vicinity of ignition sources or in potentially explosive areas. Used to detect thermal faults in electronics, to join small parts by means of cold shrinkage, to remove sticky dirt from textiles, to quick-freeze rubber and plastic and to test the function of thermostats and temperature probes.
  • Cools down to −55 °C
  • No risk of ignition or explosion, even in the presence of ignition sources
  • Does not contain any CFCs