U heeft gezocht op: Homogenisatiezakken
13  results were found
SearchResultCount:"13"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: VWR Collection
Omschrijving: Compleet assortiment zakken, steriel en voedingsgecertificeerd, voor het veilig en snel voorbereiden van vaste monsters vóór microbiologische en chemi...
Leverancier: VWR Collection
Omschrijving: Brede keuze aan PE-zakjes met een capaciteit van 80 ml tot 3500 ml
Geschikt voor alle gangbare laboratoriummixers, aangepast aan alle m...
Leverancier: NASCO FORT ATKINSON
Omschrijving: Blended, virgin polyethylene film provides exceptional clarity, and superior strength for use in homogenizer. HACCP, EPA, USDA, FDA approved for food ...
Leverancier: NASCO FORT ATKINSON
Omschrijving: Sterilised post-manufacturing and specially designed for use with homogeniser blenders. HACCP, EPA, USDA, FDA approved for food contact.
Leverancier: INTERSCIENCE
Omschrijving: BagPage® single use bags with filter membrane reduce analysis time thanks to their wide opening and markings showing where to introduce the pipette. I...
Leverancier: INTERSCIENCE
Omschrijving: The BagFilter® Pipet & Roll bags are rigid and transparent blender bags with a lateral non-woven filter for all microbiological analyses. They are ide...
Leverancier: INTERSCIENCE
Omschrijving: These non-filter sample preparation bags are ideal for homogenising samples.
Leverancier: INTERSCIENCE
Omschrijving: These lateral filter bags are with non-woven filter that are ideal for the homogenisation and automatic filtration of the samples.
Catalogus nummer: (INSC111325)
Leverancier: INTERSCIENCE
Omschrijving: BagFilter® XF is a rigid and transparent blender bag with a lateral non-woven filter for all microbiological analyses. The filtration is instantaneous...
UOM: 1 * 500 ST
Leverancier: Seward (FermionX)
Omschrijving: Stomacher®-zakken worden allemaal vervaardigd uit primaire polymeerkorrels die in een buis zijn geëxtrudeerd. De enige afdichting bevindt zich aan de ...
Leverancier: Grade Products
Omschrijving: Hoogwaardig polymeermengsel, ideaal voor laboratoriumhomogenisatoren voor voedingsmonsters.
Leverancier: Seward (FermionX)
Omschrijving: Homogenisatiezak, Simplettes®, Simplettes® (250 mm), Verpakt: 1000 in 20 (sachets of 50)
Catalogus nummer: (DELT15005)
Leverancier: DELTALAB
Omschrijving: These are sterile bags having a full size filter dividing them into two identical pouches, in which sample is inserted into one compartment. After sam...
UOM: 1 * 500 ST
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 13 of 13
no targeter for Bottom