U heeft gezocht op: Draadmanden
26  results were found
SearchResultCount:"26"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (216-2254)
Leverancier: VWR Collection
Omschrijving: Deze draadmand is gemaakt van roestvrij, elektrolytisch gepolijst roestvrij staal.
UOM: 1 * 1 ST
Leverancier: VWR Collection
Omschrijving: These wire baskets are made from rust-proof, electrolytically polished stainless steel.
Leverancier: VWR Collection
Omschrijving: These wire baskets are made from rust-proof, electrolytically polished stainless steel.
Leverancier: BOCHEM
Omschrijving: These stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
Leverancier: HUPFER METALLWERKE
Omschrijving: These fine mesh baskets, made of stainless steel, electropolished, are designed to keep small parts and accessories.
Leverancier: ELMA SCHMIDBAUER GMBH
Omschrijving: Immersion baskets, Hostalen basket, for S 30, S 30/H, Rond, b×d×h: 200×95×57 mm
Leverancier: BOCHEM
Omschrijving: Stainless steel 18/10.
Leverancier: BOCHEM
Omschrijving: Roestvrij staal.
Catalogus nummer: (142-0013)
Leverancier: ELMA SCHMIDBAUER GMBH
Omschrijving: Draadmand
UOM: 1 * 1 ST
Catalogus nummer: (BOCH10120)
Leverancier: BOCHEM
Omschrijving: These rectangular stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
UOM: 1 * 1 ST
Leverancier: HUPFER METALLWERKE
Omschrijving: Roestvrij staal:
Leverancier: RSG
Omschrijving: These electropolished wire baskets are made of stainless steel ¹⁸/₁₀.
Leverancier: HUPFER METALLWERKE
Omschrijving: Roestvrij staal.
Sale
Catalogus nummer: (HUPF8600029)
Leverancier: HUPFER METALLWERKE
Omschrijving: These wire waste paper baskets are manufactured from high-quality stainless steel, free-standing or hangable.
UOM: 1 * 1 ST
Catalogus nummer: (216-7470)
Leverancier: HUPFER METALLWERKE
Omschrijving: Stainless steel, electrolytically polished.
UOM: 1 * 1 ST
Leverancier: RSG
Omschrijving: Made from stainless steel 1.4301.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 26
no targeter for Bottom