U heeft gezocht op: Flessen met verstuiver
21  results were found
SearchResultCount:"21"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: BÃœRKLE
Omschrijving: LDPE, transparant, PP spuitopzetstuk, PE slangen, roestvrijstalen gewicht.
Leverancier: BÃœRKLE
Omschrijving: Bottle made of HDPE, spray pump made of PP and stainless steel.
Catalogus nummer: (300-0406)
Leverancier: BÃœRKLE
Omschrijving: The practical 360° spray function also allows for upside-down spraying with the Turn'n' Spray pressure sprayer, making it easy to reach typically diff...
UOM: 1 * 1 ST
Catalogus nummer: (215-4364)
Leverancier: BÃœRKLE
Omschrijving: Basic spray bottles are suitable for occasional spraying in various industries.
UOM: 1 * 1 ST
Leverancier: BÃœRKLE
Omschrijving: LDPE, doorschijnend, PP-spuitopzetstuk.
Catalogus nummer: (300-0407)
Leverancier: BÃœRKLE
Omschrijving: The food pressure sprayer has a transparent viewing window to make it easy to read the fill level. A rubber-coated ring stand ensures that the sprayer...
UOM: 1 * 1 ST
Catalogus nummer: (215-6550)
Leverancier: BÃœRKLE
Omschrijving: Media-contacting parts PP, PE, glass-fibre-reinforced PA, V2A, FKM, bottle made of HDPE.
UOM: 1 * 1 ST
Leverancier: LAMAPLAST
Omschrijving: LDPE, translucent.
Catalogus nummer: (HECH49119010)
Leverancier: GLASWARENFABRIK KARL HECHT
Omschrijving: PE spray bottles with adjustable jet.
UOM: 1 * 1 ST
Leverancier: VITLAB
Omschrijving: White or translucent made from PP.
Catalogus nummer: (KART328)
Leverancier: KARTELL
Omschrijving: HDPE/PP.
UOM: 1 * 5 ST
Leverancier: DWK Life Sciences
Omschrijving: Bottle made of white LDPE (Low Density Polyethylene).
Catalogus nummer: (141-0285)
Leverancier: WERNER & MERTZ BENELUX SA NV
Omschrijving: Foam gun for applying a stable foam carpet on horizontal and vertical surfaces. The dosage can be adjusted.
UOM: 1 * 1 ST
Catalogus nummer: (215-4419)
Leverancier: KARTELL
Omschrijving: HDPE, translucent.
UOM: 1 * 5 ST
Leverancier: MURRAY, S & CO LTD
Omschrijving: HDPE, translucent.
Leverancier: Brand
Omschrijving: HDPE, wit.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 21
no targeter for Bottom