BOD flessen
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
215-1070EA 208 EUR
POULGS748/A
BOD flessen
Glass, clear.
BOD bottles (Biological Oxygen Demand), have glass peg stoppers adjusted to leave a 'well' for water seal and a pointed cone to prevent air entrapment.
BOD bottles (Biological Oxygen Demand), have glass peg stoppers adjusted to leave a 'well' for water seal and a pointed cone to prevent air entrapment.