Bottles, narrow neck, round, HDPE, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, translucent with PP screw cap.
- Good chemical resistance
- Heat resistant up to +100 °C
- Leakproof
Ideal for storing and transporting liquids. Suitable for a wide range of applications.
Specifications
- Catalog No:
- 215-7502
- 215-7503
- 215-7511
- 215-7509
- 215-7508
- 215-7510
- 215-7506
- 215-7504
- 215-7507
- 215-7505
- 215-7501
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Environmental certification:
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
NAR Certificate of Retirement - NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
Specifications
Related Information
Product Information
- Catalog #Supplier #CapacityThreadNeck I-ØHeightExt.ØPkAvailabilityPrice
- Supplier Article Number: 2002-9125Local Article Number: NALG2002-9125Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ACT Label - Supplier Article Number: 2002-9025Local Article Number: NALG2002-9025Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ACT Label - Supplier Article Number: 2002-9050Local Article Number: NALG2002-9050Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT LabelLow Manufacturing Impact Product Transparency and Disclosure - Supplier Article Number: 2002-0001Local Article Number: NALG2002-0001Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ACT Label - Supplier Article Number: 2002-0002Local Article Number: NALG2002-0002Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ACT LabelLow Manufacturing Impact Product Transparency and Disclosure - Supplier Article Number: 2002-0004Local Article Number: NALG2002-0004Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationISO14001 (Certificate Registration No. EMS 752603), ACT Label
NAR Certificate of RetirementLow Manufacturing Impact Product Transparency and Disclosure - Supplier Article Number: 2002-0006Local Article Number: NALG2002-0006Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ACT LabelLow Manufacturing Impact Product Transparency and Disclosure - Supplier Article Number: 2002-0008Local Article Number: NALG2002-0008Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ACT Label - Supplier Article Number: 2002-0016Local Article Number: NALG2002-0016Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ACT Label - Supplier Article Number: 2002-9016Local Article Number: NALG2002-9016Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Supplier Article Number: 2002-0032Local Article Number: NALG2002-0032Specifications:
Low Manufacturing Impact Product Transparency and Disclosure More
Specifications
Environmentally PreferableLow Manufacturing Impact, Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ACT LabelLow Manufacturing Impact Product Transparency and Disclosure