U heeft gezocht op: Bakken voor buisjes
7  results were found
SearchResultCount:"7"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (211-0370)
Leverancier: VWR Collection
Omschrijving: PP, met transparant deksel
UOM: 1 * 5 ST
Leverancier: Simport Scientific
Omschrijving: Cover and base made of polystyrene.
Leverancier: Simport Scientific
Omschrijving: Cover is made of PS and base made of high impact PS.
Catalogus nummer: (HEATHS120018)
Leverancier: HEATHROW
Omschrijving: Expanding storage rack reduce sample handling time and the need for several racks when working with samples for cold temperature storage. The opening ...
UOM: 1 * 40 ST
Leverancier: GLW
Omschrijving: Styro racks are made of heavy expanded polystyrene which are used in freezer down to -90 °C. System box is made of cardboard and is designed to secure...
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Omschrijving: Box in bright colours made of PP for cell culture tubes of 15 or 50 ml (Falcon). Two grids guarantee a safe stand of the tubes in the upper as well as...
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Omschrijving: Boxes made of transparent glass-clear polystyrene. Suitable for variable applications.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 7 of 7
no targeter for Bottom