Emmers, roestvrij staal
Leverancier: Coldstream
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COLDBUC4010GEA 224 EUR
COLDBUC4010G COLDBUC4015G COLDBUC4012G
Emmers, roestvrij staal
Emmers
Stainless steel, 304 grade.
  • Very good chemical resistance, very good thermal resistance
  • High quality, very stable
  • An optional lid is available which is extended to cover the spout
These buckets are polished throughout, graduated in 1 litre intervals and come complete with swing handle. Skirtbands are standard. They have crevice and pit free interiors and are polished throughout.