Netherlands
Order Entry
VWR®, Scientific Calculator
VWR®, Scientific Calculator
Catalog # 818-0062
Supplier:  VWR Collection
undefined
VWR®, Scientific Calculator
Catalog # 818-0062
Supplier:  VWR Collection
Local Article Number: VWRI818-0062
Price
45,80 €
Log in for availability
Unit of Measure 1 * 1 ST
Specifications
  • Description:
    Scientific calculator
  • W×D×H:
    76×13×127 mm
  • Pk:
    1
Specifications
About this item
Convenient pocket size calculator can handle 99% of all routine and other calculations in the laboratory. Provides immediate, exact results for a wide range of applications: Conversions, scientific calculations, statistical operations, arithmetic calculations and similar laboratory work. Display: 10 characters with two-digit exponent.
  • Clear display with large, bright numbers and ergonomic keys
  • Three key memory and memory protection when the calculator is switched off
  • Automatic switch-off to preserve batteries
Functions: Over 50 functions for trigonometry calculations, logarithms, exponentials and roots, inverse calculations, factoring, percentages, hexadecimal calculations, probability generator and conversion from degrees to decimal degrees. The statistics function provides standard deviation, averages, sum of squares, sum of data and much more.
Delivery information: Batteries included.