Laboratory coats, Tyvek® 500
Supplier: DuPont
Certificates
About this item
These garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
- Fabric is ultra-low linting
- With collar and stitched internal seams
- Anti-static treatment (EN 1149-5) on both sides
- PL30 with straight cuffs, PL309 + PL30NP with elasticated cuffs (not tunnelled)
- Available with zip or press stud closure, with or without pockets
Typical applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up.
Colour: White
Cat. I
Certifications: (EU) 2016/425; partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents).
Packaging: 50 per box, bulk packed.
Specifications
- Catalog No:
- 113-0467
- 113-0473
- 113-0474
- 113-0475
- 113-0472
- 113-3882
- 113-0471
- 113-0477
- 113-0470
- 113-0478
- 113-0476
- 113-0468
- 113-0469
- Model:
- PL30, 3 pockets
- PL309, 2 pockets
- PL309, 2 pockets
- PL30NP
- PL309, 2 pockets
- PL309, 2 pockets
- PL309, 2 pockets
- PL30NP
- PL30, 3 pockets
- PL30NP
- PL30NP
- PL30, 3 pockets
- PL30, 3 pockets
- Closure type:
- Press studs / snaps
- Zip closure
- Zip closure
- Press studs / snaps
- Zip closure
- Zip closure
- Zip closure
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- With pockets
- With pockets
- With pockets
- Without pockets
- With pockets
- With pockets
- With pockets
- Without pockets
- With pockets
- Without pockets
- Without pockets
- With pockets
- With pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Chemical protection, Laboratory
- Weight:
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²
Specifications
PL30: press-stud closure, 3 pockets
Product Information
- Catalog #Supplier #ColourClothing SizePkAvailabilityPrice
- Supplier Article Number: TYPL30SWHLG53PLocal Article Number: DUPOTYPL30SWHLG53PSpecifications:
More
Specifications
ModelPL30, 3 pocketsEnvironmentally PreferableClosure typePress studs / snapsPocketsWith pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWH2X53PLocal Article Number: DUPOTYPL30SWH2X53PSpecifications:
More
Specifications
ModelPL30, 3 pocketsEnvironmentally PreferableClosure typePress studs / snapsPocketsWith pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWHMD53PLocal Article Number: DUPOTYPL30SWHMD53PSpecifications:
More
Specifications
ModelPL30, 3 pocketsEnvironmentally PreferableClosure typePress studs / snapsPocketsWith pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWHXL53PLocal Article Number: DUPOTYPL30SWHXL53PSpecifications:
More
Specifications
ModelPL30, 3 pocketsEnvironmentally PreferableClosure typePress studs / snapsPocketsWith pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m²
PL309: Zip closure, 2 pockets
Product Information
- Catalog #Supplier #ColourClothing SizePkAvailabilityPrice
- Supplier Article Number: TYPL30SWHSM005Local Article Number: DUPOTYPL30SWHSM005Specifications:
More
Specifications
ModelPL309, 2 pocketsEnvironmentally PreferableClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWHLG52PLocal Article Number: DUPOTYPL30SWHLG52PSpecifications:
More
Specifications
ModelPL309, 2 pocketsEnvironmentally PreferableClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWHXL52PLocal Article Number: DUPOTYPL30SWHXL52PSpecifications:
More
Specifications
ModelPL309, 2 pocketsEnvironmentally PreferableClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWH2X52PLocal Article Number: DUPOTYPL30SWH2X52PSpecifications:
More
Specifications
ModelPL309, 2 pocketsEnvironmentally PreferableClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWHMD52PLocal Article Number: DUPOTYPL30SWHMD52PSpecifications:
More
Specifications
ModelPL309, 2 pocketsEnvironmentally PreferableClosure typeZip closurePocketsWith pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m²
PL30NP: Press-stud closure, no pockets
Product Information
- Catalog #Supplier #ColourClothing SizePkAvailabilityPrice
- Supplier Article Number: TYPL30SWHLG5NPLocal Article Number: DUPOTYPL30SWHLG5NPSpecifications:
More
Specifications
ModelPL30NPEnvironmentally PreferableClosure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWHXL5NPLocal Article Number: DUPOTYPL30SWHXL5NPSpecifications:
More
Specifications
ModelPL30NPEnvironmentally PreferableClosure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWHMD5NPLocal Article Number: DUPOTYPL30SWHMD5NPSpecifications:
More
Specifications
ModelPL30NPEnvironmentally PreferableClosure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m² - Supplier Article Number: TYPL30SWH2X5NPLocal Article Number: DUPOTYPL30SWH2X5NPSpecifications:
More
Specifications
ModelPL30NPEnvironmentally PreferableClosure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationChemical protection, LaboratoryWeight41 g/m²