Overalls, Tritex® pro, SMS
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Certificates
About this item
This overall made from three ply SMS fabric is robust, breathable and anti-static. It offers comfort in all work areas.
- Hood with elastic tie
- Zip covering strip
- Elasticated waist, sleeves and legs
Suitable for production and bottling activities in the food, pharmaceutical or cosmetic industries as well as for production and processing in the chemical and veterinarian industries etc.
Colour: White.
Certifications: CE 0624, Cat. III, Type 5/6
Specifications
- Catalog No:
- 113-0656
- 113-0655
- 113-0654
- Anti-static:
- +
- +
- +
- Garment application:
- Laboratory, Food
- Laboratory, Food
- Laboratory, Food
Specifications
Product Information
- Catalog #Supplier #ColourClothing SizePkAvailabilityPrice
- Supplier Article Number: 30753-90Local Article Number: ZVGA30753-90Specifications:
More
Specifications
Environmentally PreferableAnti-static+GenderGarment applicationLaboratory, Food - Supplier Article Number: 30754-90Local Article Number: ZVGA30754-90Specifications:
More
Specifications
Environmentally PreferableAnti-static+GenderGarment applicationLaboratory, Food - Supplier Article Number: 30755-90Local Article Number: ZVGA30755-90Specifications:
More
Specifications
Environmentally PreferableAnti-static+GenderGarment applicationLaboratory, Food