About this item
PRP-1 polymeric HPLC columns are designed for broad ranging uses, similar to C8 or C18 HPLC materials. They combine the best characteristics of silica-based and polymeric columns resulting in highly inert and long-lasting columns. These columns are ideal for applications that require a high degree of column to column and batch to batch consistency (quality assurance, clinical trials). Unlike C8 or C18 columns, PRP-1 has no stationary phase coating. Since there is no stationary phase to hydrolyse, the column maintains its performance characteristics longer than many C8 or C18 columns.
- Made entirely of polystyrene-divinylbenzene, there are no silanol groups to cause sample loss
- Cross-linked to prevent shrinkage or swelling
Quality assurance as well as research and development laboratories find that PRP-1 columns last significantly longer than silica-based C8 or C18 columns. This is particularly true for difficult analyses such as high pH samples (pH 8-13), labile or reactive samples (irreversible adsorption), high aqueous purifications (80-100% water), and separations with ion pairing reagents.
Specifications
- Catalog No:
- 554-0233
- 554-0431
- 554-1313
- 554-0433
- 554-0466
- 554-0284
- 554-0289
- 554-0484
- 554-0290
- 554-1308
- 554-0286
- 554-1260
- 554-0257
- 554-0230
- 554-0232
- 554-0231
- 554-0203
- 554-1494
- 554-1262
- 554-0285
- 554-1221
- 554-1222
- 554-0245
- 554-0258
- 554-1259
- 554-0214
- 554-0294
- 554-0468
- 554-0229
- 554-1241
- 554-0211
- 554-0213
- 554-2531
- 554-0234
- 554-0295
- 554-2536
- 554-1240
- 554-5698
- 554-2537
- 554-1312
- 554-0202
- 554-5700
- Max. pressure:
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- pH range:
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- USP code:
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
Specifications
Product Information
- Catalog #Supplier #Column FormatPhaseColumn hardwareParticle SizeØ int.AvailabilityPrice
- Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-253779355Semi-preparativePRP-1Stainless steel5 µm10,00 mmLog in for availability1 * 1 ST2 260,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-253179154Semi-preparativePRP-1Stainless steel5 µm21,20 mmLog in for availability1 * 1 ST3 430,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-029579531Semi-preparativePRP-1Stainless steel7 µm10,00 mmLog in for availability1 * 1 ST2 260,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-149479367Semi-preparativePRP-1Stainless steel7 µm10,00 mmLog in for availability1 * 1 ST1 820,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-020379352Semi-preparativePRP-1Stainless steel7 µm21,20 mmLog in for availability1 * 1 ST6 160,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-028579495AnalyticalPRP-1Stainless steel10 µm7,00 mmLog in for availability1 * 1 ST1 560,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-023279426AnalyticalPRP-1Stainless steel10 µm7,00 mmLog in for availability1 * 1 ST1 270,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-028979499Semi-preparativePRP-1Stainless steel10 µm10,00 mmLog in for availability1 * 1 ST1 690,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-028679496Semi-preparativePRP-1Stainless steel10 µm10,00 mmLog in for availability1 * 1 ST1 950,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-025779478Semi-preparativePRP-1Stainless steel10 µm21,20 mmLog in for availability1 * 1 ST5 890,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-131279567PreparativePRP-1Stainless steel10 µm50,80 mmLog in for availability1 * 1 ST18 840,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-028479493PreparativePRP-1Stainless steel12 - 20 µm50,80 mmLog in for availability1 * 1 ST20 990,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-029079525PreparativePRP-1Stainless steel12 - 20 µm101,60 mmLog in for availability1 * 1 ST34 980,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-023479428Semi-preparativePRP-1Stainless steel12 - 20 µm21,20 mmLog in for availability1 * 1 ST5 600,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21 - 554-253679229PreparativePRP-1Stainless steel12 - 20 µm30,00 mmLog in for availability1 * 1 ST6 320,00 €Specifications:
More
Specifications
Environmentally PreferableMax. pressure5000 psipH range1 - 13USP codeL21
Analytical guard columns
Product Information
- Catalog #Supplier #Column FormatPhaseColumn hardwareParticle SizeØ int.AvailabilityPrice
- Analytical guard cartridge replacement cartridgesAnalytical guard cartridge replacement cartridges554-126079445Guard columnPRP-1Stainless steel10 µm2,30 mmLog in for availability1 * 5 ST364,00 €Specifications:
Specifications
Environmentally PreferableMax. pressurepH rangeUSP code - Analytical guard columnAnalytical guard column554-569879286Guard columnPRP-1Stainless steel10 µm2,10 mmLog in for availability1 * 1 ST453,00 €Specifications:
Specifications
Environmentally PreferableMax. pressurepH rangeUSP code - Analytical guard cartridge starter kit (1 holder, 2 cartridges)Analytical guard cartridge starter kit (1 holder, 2 cartridges)554-126279447Guard columnPRP-1Stainless steel10 µm2,30 mmLog in for availability1 * 1 ST330,00 €Specifications:
Specifications
Environmentally PreferableMax. pressurepH rangeUSP code - Analytical guard cartridge replacement cartridgesAnalytical guard cartridge replacement cartridges554-124179318Guard columnPRP-1PEEK10 µm3,00 mmLog in for availability1 * 5 ST448,00 €Specifications:
Specifications
Environmentally PreferableMax. pressurepH rangeUSP code - Analytical guard cartridge starter kit (1 holder, 2 cartridges)Analytical guard cartridge starter kit (1 holder, 2 cartridges)554-124079317Guard columnPRP-1PEEK10 µm3,00 mmLog in for availability1 * 1 ST413,00 €Specifications:
Specifications
Environmentally PreferableMax. pressurepH rangeUSP code
Semi-preparative/preparative guard columns
Product Information
- Catalog #Supplier #Column FormatPhaseColumn hardwareParticle SizeØ int.AvailabilityPrice
- Semiprep/Preparative guard column starter kit (1 holder, 1 cartridge)Semiprep/Preparative guard column starter kit (1 holder, 1 cartridge)554-122179121Guard columnPRP-1Stainless steel12 - 20 µm4,60 mmLog in for availability1 * 1 ST526,00 €Specifications:
Specifications
Environmentally PreferableMax. pressurepH rangeUSP code - Semiprep/Preparative guard column replacement cartridgesSemiprep/Preparative guard column replacement cartridges554-122279122Guard columnPRP-1Stainless steel12 - 20 µm4,60 mmLog in for availability1 * 2 ST386,00 €Specifications:
Specifications
Environmentally PreferableMax. pressurepH rangeUSP code - Semiprep/Preparative guard columnSemiprep/Preparative guard column554-570079912Guard columnPRP-1Stainless steel12 - 20 µm4,60 mmLog in for availability1 * 1 ST453,00 €Specifications:
Specifications
Environmentally PreferableMax. pressurepH rangeUSP code