U heeft gezocht op: Koelelementen
10  results were found
SearchResultCount:"10"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: VWR Collection
Omschrijving: Met gel gevulde plastic koffer.
Leverancier: VWR Collection
Omschrijving: Gelvulling.
Leverancier: VWR Collection
Omschrijving: Halfhard schuim.
Leverancier: Sonoco Thermosafe
Omschrijving: Polar Pack® refrigerants and Utek® phase change materials provide thermal solutions for shipping temperature critical packages during transit. Availab...
Catalogus nummer: (SCATFPP24)
Leverancier: Sonoco Thermosafe
Omschrijving: PolarPack® gel brick coolant pack refrigerants provide thermal solutions for temperature critical packages during transit and are an alternative to we...
UOM: 1 * 768 ST
Catalogus nummer: (DLTTA090007)
Leverancier: DELTA T
Omschrijving: 37 C Accus maintain a temperature between 36 to 38 °C.
UOM: 1 * 1 ST
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Omschrijving: The inserts are simply mounted in the special groove.
Leverancier: HONIGBERG, J.D. INTERNATIONAL
Omschrijving: Heavy, metal-coated PE film filled with special gel cooling pad
Sale
Leverancier: CAMPING GAZ INTERNATIONAL
Omschrijving: This cooling pack is ideal for keeping drinks, food or medicines cool for longer in a passive hard or soft cooler. Its components include water, carbo...
Catalogus nummer: (216-7015)
Leverancier: HONIGBERG, J.D. INTERNATIONAL
Omschrijving: For cooling, shipping and cold storage. Ice replacement in small plastic bags, for embedding samples and other frozen food. Due to their high flexibil...
UOM: 1 * 1.500 ST
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 10 of 10
no targeter for Bottom