U heeft gezocht op: Kroezen
67  results were found
SearchResultCount:"67"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: VWR Collection
Omschrijving: Porselein, van binnen en van buiten geglazuurd, behalve de bodem.
Leverancier: VWR Collection
Omschrijving: Porselein, met poreuze bodem.
Leverancier: VWR Collection
Omschrijving: Isostatisch gegoten in zuiver PTFE, met FDA goedgekeurd.
Leverancier: VWR Collection
Omschrijving: Smeltkroezen uit roestvrij staal 304.
Leverancier: ROBU GLASFILTER
Omschrijving: Filterkroezen met geïntegreerde filterhouder uit gesinterd glas. Borosilicaatglas 3.3.
Leverancier: ROBU GLASFILTER
Omschrijving: These filltration crucibles are suitable for crude fibre extraction.
Leverancier: TECHNICAL QUARTZ SOLUTIONS GMB
Omschrijving: These crucibles are made from fused quartz, translucent, glazed.
Leverancier: DWK Life Sciences
Omschrijving: These filtration crucibles are made from clear DURAN® borosilicate glass 3.3.
Leverancier: USBECK Laborgeräte
Omschrijving: These crucibles are made from iron.
Leverancier: TECHNICAL QUARTZ SOLUTIONS GMB
Omschrijving: These crucibles are made from fused quartz, translucent, glazed.
Leverancier: Thermo Scientific
Omschrijving: These crucibles are manufactured from unpurified extruded graphite.
Leverancier: Thermo Scientific
Omschrijving: These crucibles are manufactured from glassy carbon.
Leverancier: Thermo Scientific
Omschrijving: These crucibles are manufactured from nickel. Available in high form, low form and straight wall types.
Leverancier: TECHNICAL QUARTZ SOLUTIONS GMB
Omschrijving: Quartz, transparent, glazed.
Leverancier: Thermo Scientific
Omschrijving: These crucibles are manufactured from yttria stabilised zirconium oxide. They display superior characteristics both at elevated temperature and at hig...
Leverancier: Thermo Scientific
Omschrijving: These crucibles are manufactured from zirconium. Available in low form and straight wall types.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 67
no targeter for Bottom