Integrated cup with adulteration test for urine
Leverancier: SureScreen Diagnostics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-1268EA 465 EUR
SUREGINCUP10DSA
Integrated cup with adulteration test for urine
This collection cup is a closed test system for when performance matters. The cup features an integrated multipanel, temperature strip, evidentially secure isolation chamber, and adulteration strip. Our multipanel fitted to a clever urine cup that isolates the urine from the test. Coupled with an adulteration test strip, the cup ensures a fresh, clean sample. A key supplied allows just the right amount of urine to the test when used, leaving the rest isolated for confirmation if required. The adulteration strip on the new integrated cup comes in three varieties: A parameters - Oxidants, Specific Gravity, Ph; B parameters - Nitrite, Glutaraldehyde, Creatinine, both A and B Parameters together on the multiline panel.
  • Sample can be directly collected
  • No mess
  • No smells