Pallflex® filters
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Versatile filters uniquely suited for a broad range of air monitoring applications.
- Can be used for high temperature applications
- Ideal for diesel and other emissions testing, required by environmental regulatory agencies
Tissuquartz™: Pure quartz, no binder. Aerosol retention: 99,9%
Fiberfilm™: Heat resistant borosilicate glass fibres coated with fluorocarbon (TFE). Aerosol retention: 96,4%
Emfab™: Borosilicate glass microfibres reinforced with woven glass cloth and bonded with PTFE. Aerosol retention: 99,95%
* non heat treated
Specifications
- Catalog No:
- 513-0034
- 516-8015
- 516-7992
- 513-0029
- 516-8014
- 516-7991
- PALL7202D
- 513-0028
- 516-7995
- 516-7985
- 513-0035
- 516-8008
- 516-8013
- 516-7996
- PALLXQ250ETDS0150
- Grade:
- Emfab™ TX40HI20WW
- Emfab™ TX40HI20WW
- Tissuquartz™
- Tissuquartz™
- Emfab™ TX40HI20WW
- Tissuquartz™
- Tissuquartz™
- Tissuquartz™
- Tissuquartz™
- Tissuquartz™
- Emfab™ TX40HI20WW
- Emfab™ TX40HI20WW
- Emfab™ TX40HI20WW
- Tissuquartz™
- Tissuquartz™
- Weight:
- 5 mg/cm²
- 5 mg/cm²
- 5,8 mg/cm²
- 5.8 mg/cm²
- 5 mg/cm²
- 5,8 mg/cm²
- 5.8 mg/cm²
- 5,8 mg/cm²
- 5,8 mg/cm²
- 5 mg/cm²
- 5 mg/cm²
- 5 mg/cm²
- 5,8 mg/cm²
- 5,8 mg/cm²
- Thickness:
- 178 µm
- 178 µm
- 432 µm
- 432 µm
- 178 µm
- 432 µm
- 432 µm
- 432 µm
- 432 µm
- 178 µm
- 178 µm
- 178 µm
- 432 µm
- 432 µm
- Water flow rate:
- 32 l/min/cm² (0,35 bar)
- 32 l/min/cm² (0,35 bar)
- 220 l/min/cm² (0,35 bar)
- 220 l/min/cm² (0,35 bar)
- 32 l/min/cm² (0,35 bar)
- 220 l/min/cm² (0,35 bar)
- 220 l/min/cm² (0,35 bar)
- 220 l/min/cm² (0,35 bar)
- 220 l/min/cm² (0,35 bar)
- 32 l/min/cm² (0,35 bar)
- 32 l/min/cm² (0,35 bar)
- 32 l/min/cm² (0,35 bar)
- 220 l/min/cm² (0,35 bar)
- 220 l/min/cm² (0,35 bar)
- Airflow rate:
- 68 l/min/cm² (0.7 bar)
- 68 l/min/cm² (0,7 bar)
- 73 l/min/cm² (0,7 bar)
- 73 l/min/cm² (0.7 bar)
- 68 l/min/cm² (0,7 bar)
- 73 l/min/cm² (0,7 bar)
- 73 l/min/cm² (0,7 bar)
- 73 l/min/cm² (0.7 bar)
- 73 l/min/cm² (0,7 bar)
- 73 l/min/cm² (0,7 bar)
- 68 l/min/cm² (0.7 bar)
- 68 l/min/cm² (0,7 bar)
- 68 l/min/cm² (0,7 bar)
- 73 l/min/cm² (0,7 bar)
- 73 l/min/cm² (0,7 bar)
- Max. temperature:
- 260 °C
- 260 °C
- 1093 °C
- 1093 °C
- 260 °C
- 1093 °C
- 1093 °C
- 1093 °C
- 1093 °C
- 1093 °C
- 260 °C
- 260 °C
- 260 °C
- 1093 °C
- 1093 °C
Specifications
Related Information
Product Information
- Catalog #Supplier #GradeSizePkAvailabilityPrice
- Specifications:
More
Specifications
GradeTissuquartz™Environmentally PreferableWeight5,8 mg/cm²Thickness432 µmWater flow rate220 l/min/cm² (0,35 bar)Airflow rate73 l/min/cm² (0,7 bar)Max. temperature1093 °C - Specifications:
More
Specifications
GradeTissuquartz™Environmentally PreferableWeight5,8 mg/cm²Thickness432 µmWater flow rate220 l/min/cm² (0,35 bar)Airflow rate73 l/min/cm² (0,7 bar)Max. temperature1093 °C - Specifications:
More
Specifications
GradeTissuquartz™Environmentally PreferableWeight5,8 mg/cm²Thickness432 µmWater flow rate220 l/min/cm² (0,35 bar)Airflow rate73 l/min/cm² (0,7 bar)Max. temperature1093 °C - Specifications:
More
Specifications
GradeTissuquartz™Environmentally PreferableWeightThicknessWater flow rateAirflow rate73 l/min/cm² (0,7 bar)Max. temperature1093 °C - Specifications:
More
Specifications
GradeTissuquartz™Environmentally PreferableWeight5.8 mg/cm²Thickness432 µmWater flow rate220 l/min/cm² (0,35 bar)Airflow rate73 l/min/cm² (0.7 bar)Max. temperature1093 °C - Specifications:
More
Specifications
GradeTissuquartz™Environmentally PreferableWeight5,8 mg/cm²Thickness432 µmWater flow rate220 l/min/cm² (0,35 bar)Airflow rate73 l/min/cm² (0,7 bar)Max. temperature1093 °C - Specifications:
More
Specifications
GradeTissuquartz™Environmentally PreferableWeight5.8 mg/cm²Thickness432 µmWater flow rate220 l/min/cm² (0,35 bar)Airflow rate73 l/min/cm² (0.7 bar)Max. temperature1093 °C - Specifications:
More
Specifications
GradeTissuquartz™Environmentally PreferableWeight5,8 mg/cm²Thickness432 µmWater flow rate220 l/min/cm² (0,35 bar)Airflow rate73 l/min/cm² (0,7 bar)Max. temperature1093 °C - Specifications:
More
Specifications
GradeTissuquartz™Environmentally PreferableWeight5,8 mg/cm²Thickness432 µmWater flow rate220 l/min/cm² (0,35 bar)Airflow rate73 l/min/cm² (0,7 bar)Max. temperature1093 °C - Specifications:
More
Specifications
GradeEmfab™ TX40HI20WWEnvironmentally PreferableWeight5 mg/cm²Thickness178 µmWater flow rate32 l/min/cm² (0,35 bar)Airflow rate68 l/min/cm² (0.7 bar)Max. temperature260 °C - Specifications:
More
Specifications
GradeEmfab™ TX40HI20WWEnvironmentally PreferableWeight5 mg/cm²Thickness178 µmWater flow rate32 l/min/cm² (0,35 bar)Airflow rate68 l/min/cm² (0,7 bar)Max. temperature260 °C - Specifications:
More
Specifications
GradeEmfab™ TX40HI20WWEnvironmentally PreferableWeight5 mg/cm²Thickness178 µmWater flow rate32 l/min/cm² (0,35 bar)Airflow rate68 l/min/cm² (0.7 bar)Max. temperature260 °C - Specifications:
More
Specifications
GradeEmfab™ TX40HI20WWEnvironmentally PreferableWeight5 mg/cm²Thickness178 µmWater flow rate32 l/min/cm² (0,35 bar)Airflow rate68 l/min/cm² (0,7 bar)Max. temperature260 °C - Specifications:
More
Specifications
GradeEmfab™ TX40HI20WWEnvironmentally PreferableWeight5 mg/cm²Thickness178 µmWater flow rate32 l/min/cm² (0,35 bar)Airflow rate68 l/min/cm² (0,7 bar)Max. temperature260 °C - Specifications:
More
Specifications
GradeEmfab™ TX40HI20WWEnvironmentally PreferableWeight5 mg/cm²Thickness178 µmWater flow rate32 l/min/cm² (0,35 bar)Airflow rate68 l/min/cm² (0,7 bar)Max. temperature260 °C