Filtertrechters, micro, VitraPOR®
Leverancier: ROBU
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VitraPOR®
511-0258EA 132 EUR
511-0258 511-0257 511-0385 511-0256 ROBU27201 511-0274 511-0384 511-0290 ROBU27081 511-0291
Filtertrechters, micro, VitraPOR®
Trechters Filtertrechters Microfiltertrechters
Borosilicaatglas 3.3.
  • For filtration of very small volumes
  • Excellent temperature resistance
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
Met geïntegreerde filterhouder uit gesinterd glas.