Netherlands
Order Entry
E-Z Load™ Histosette® II M482 & M483, E-Z Load™ QuickLoad™ M485SL & M486SL
E-Z Load™ Histosette® II M482 & M483, E-Z Load™ QuickLoad™ M485SL & M486SL
Supplier:  Simport Scientific
Certificates
undefined
E-Z Load™ Histosette® II M482 & M483, E-Z Load™ QuickLoad™ M485SL & M486SL
Supplier:  Simport Scientific
Tissue cassette bases M482
Product Information
  • Catalog #
    Supplier #
    Colour
    Pk
    Availability
    Price
  • 720-0850
    M482-4BA
    Green
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0848
    M482-3BA
    Pink
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0856
    M482-7BA
    Pale orange
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0840
    M482-10BA
    Lilac
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0854
    M482-6BA
    Blue
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0852
    M482-5BA
    Yellow
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0846
    M482-2BA
    White
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0842
    M482-11BA
    Orange
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0844
    M482-12BA
    Turquoise
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0860
    M482-9BA
    Grey
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0858
    M482-8BA
    Cream
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 271,00 €
    271,00 €
    1 * 1.000 ST
       271,00 €
    Specifications:
    Specifications
    Environmentally Preferable
Tissue cassette covers M482
Product Information
  • Catalog #
    Supplier #
    Colour
    Pk
    Availability
    Price
  • 720-0849
    M482-3LI
    Pink
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0847
    M482-2LI
    White
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0851
    M482-4LI
    Green
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0841
    M482-10LI
    Lilac
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0845
    M482-12LI
    Turquoise
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0843
    M482-11LI
    Orange
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0855
    M482-6LI
    Blue
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0861
    M482-9LI
    Grey
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0859
    M482-8LI
    Tan
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0857
    M482-7LI
    Pale orange
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0853
    M482-5LI
    Yellow
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,30 €
    80,30 €
    1 * 1.000 ST
       80,30 €
    Specifications:
    Specifications
    Environmentally Preferable
Biopsy cassette bases M483
Product Information
  • Catalog #
    Supplier #
    Colour
    Pk
    Availability
    Price
  • 720-0882
    M483-9BA
    Grey
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0872
    M483-4BA
    Green
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0880
    M483-8BA
    Tan
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0878
    M483-7BA
    Pale orange
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0876
    M483-6BA
    Blue
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0874
    M483-5BA
    Yellow
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0870
    M483-3BA
    Pink
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0868
    M483-2BA
    White
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0866
    M483-12BA
    Turquoise
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0864
    M483-11BA
    Orange
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0862
    M483-10BA
    Lilac
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 303,00 €
    303,00 €
    1 * 1.000 ST
       303,00 €
    Specifications:
    Specifications
    Environmentally Preferable
Biopsy cassette covers M483
Product Information
  • Catalog #
    Supplier #
    Colour
    Pk
    Availability
    Price
  • 720-0883
    M483-9LI
    Grey
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0881
    M483-8LI
    Tan
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0875
    M483-5LI
    Yellow
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0871
    M483-3LI
    Pink
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0879
    M483-7LI
    Pale orange
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0873
    M483-4LI
    Green
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0869
    M483-2LI
    White
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0867
    M483-12LI
    Turquoise
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0865
    M483-11LI
    Orange
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0877
    M483-6LI
    Blue
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-0863
    M483-10LI
    Lilac
    1.000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 90,80 €
    90,80 €
    1 * 1.000 ST
       90,80 €
    Specifications:
    Specifications
    Environmentally Preferable
Tissue cassettes (with cover) in QuickLoad™ sleeves, M485SL
Product Information
  • Catalog #
    Supplier #
    Colour
    Pk
    Availability
    Price
  • 720-1213
    M485-5SL
    Yellow
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 151,00 €
    151,00 €
    1 * 750 ST
       151,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1214
    M485-6SL
    Blue
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 151,00 €
    151,00 €
    1 * 750 ST
       151,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1211
    M485-3SL
    Pink
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 151,00 €
    151,00 €
    1 * 750 ST
       151,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1212
    M485-4SL
    Green
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 151,00 €
    151,00 €
    1 * 750 ST
       151,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1210
    M485-2SL
    White
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 151,00 €
    151,00 €
    1 * 750 ST
       151,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1209
    M485-12SL
    Turquoise
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 151,00 €
    151,00 €
    1 * 750 ST
       151,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1207
    M485-10SL
    Lilac
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 151,00 €
    151,00 €
    1 * 750 ST
       151,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1208
    M485-11SL
    Orange
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 151,00 €
    151,00 €
    1 * 750 ST
       151,00 €
    Specifications:
    Specifications
    Environmentally Preferable
Biopsy cassettes (with cover) in QuickLoad™ sleeves, M486SL
Product Information
  • Catalog #
    Supplier #
    Colour
    Pk
    Availability
    Price
  • 720-1222
    M486-6SL
    Blue
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 217,00 €
    217,00 €
    1 * 750 ST
       217,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1219
    M486-3SL
    Pink
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 217,00 €
    217,00 €
    1 * 750 ST
       217,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1220
    M486-4SL
    Green
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 217,00 €
    217,00 €
    1 * 750 ST
       217,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1218
    M486-2SL
    White
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 217,00 €
    217,00 €
    1 * 750 ST
       217,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1216
    M486-11SL
    Orange
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 217,00 €
    217,00 €
    1 * 750 ST
       217,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1217
    M486-12SL
    Turquoise
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 217,00 €
    217,00 €
    1 * 750 ST
       217,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1215
    M486-10SL
    Lilac
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 217,00 €
    217,00 €
    1 * 750 ST
       217,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • 720-1221
    M486-5SL
    Yellow
    750
    Log in for availability
    Log in for availability
    1 * 750 ST
    1 * 750 ST - 217,00 €
    217,00 €
    1 * 750 ST
       217,00 €
    Specifications:
    Specifications
    Environmentally Preferable