Drums, total opening, with screw lids
Supplier: CURTEC
Certificates
About this item
Drums, HDPE, white, with red screw lids or HDPE brown with beige screw lids and EPDM sealing gasket.
- Airtight and watertight
- Total opening containers
- 100% plastic, free of heavy metals
Stackable wide mouth kegs with lid, tamper-resistant seal and predetermined breaking point. Made from food grade material and UN approved for solids and pastes.
CurTec has introduced a material clock. The material clock is a configurable clock that shows the materials used. Each letter of the clock corresponds to a specific material. These letters, together with the respective materials, are described in the UN certificate. The material clock can be placed on the bottom of the container.
White drum, red lid
Product Information
- Catalog #Supplier #CapacityExt.ØØ openingHeightPkAvailabilityPrice
- Supplier Article Number: 6945-61-046Local Article Number: CURT6945-61-046Specifications:
Accessories
Specifications
Environmentally Preferable
White drum, red lid, with handgrips
Product Information
- Catalog #Supplier #CapacityExt.ØØ openingHeightPkAvailabilityPrice
- Supplier Article Number: 6950-91-046Local Article Number: CURT6950-91-046Specifications:
Accessories
Specifications
Environmentally Preferable - Supplier Article Number: 6947-91-046Local Article Number: CURT6947-91-046Specifications:
Accessories
Specifications
Environmentally Preferable - Supplier Article Number: 6994-91-046Local Article Number: CURT6994-91-046Specifications:
Accessories
Specifications
Environmentally Preferable
Brown drum, beige lid, with handgrips
Product Information
- Catalog #Supplier #CapacityExt.ØØ openingHeightPkAvailabilityPrice
- Supplier Article Number: 6942-91-971Local Article Number: CURT6942-91-971Specifications:
Accessories
Specifications
Environmentally Preferable - Supplier Article Number: 6943-91-971Local Article Number: CURT6943-91-971Specifications:
Accessories
Specifications
Environmentally Preferable