Netherlands
Order Entry
Cryo-Babies® and Cryo-Tags® cryo label rolls
Cryo-Babies® and Cryo-Tags® cryo label rolls
Certificates
undefined
Cryo-Babies® and Cryo-Tags® cryo label rolls
Product Information
  • Catalog #
    Supplier #
    Material
    Printing method
    Colour
    Printer model
    Dimensions
    Availability
    Price
  • Crypo labels, Cryo-Babies for 0,5 ml tubes
    Crypo labels, Cryo-Babies for 0,5 ml tubes
    817-0105
    CRY-0500
    PVC
    White
    24×13 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 89,20 €
    89,20 €
    1 * 1.000 ST
       89,20 €
    Supplier Article Number: CRY-0500
    Local Article Number: DIVECRY-0500
    Specifications:
    Specifications
    Environmentally Preferable
  • Crypo labels, Cryo-Babies for 1,5 - 2,0 ml tubes
    Crypo labels, Cryo-Babies for 1,5 - 2,0 ml tubes
    817-2131
    CRY-1000
    PVC
    White
    33×13 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 89,20 €
    89,20 €
    1 * 1.000 ST
       89,20 €
    Supplier Article Number: CRY-1000
    Local Article Number: DIVECRY-1000
    Specifications:
    Specifications
    Environmentally Preferable
  • Cryo labels, F2894
    Cryo labels, F2894
    DIVECRY-2000
    CRY-2000
    PVC
    White
    38×19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 89,20 €
    89,20 €
    1 * 1.000 ST
       89,20 €
    Supplier Article Number: CRY-2000
    Local Article Number: DIVECRY-2000
    Specifications:
    Specifications
    Environmentally Preferable