Glass bottom microplates, SensoPlate™
Leverancier: Greiner bio-one
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SensoPlate™ SensoPlate™ Plus
738-0031EA 598 EUR
738-0031 GREI655896_16 738-0504 738-0027 738-0104 738-0105 GREI655891_16
Glass bottom microplates, SensoPlate™
Microplaten HTS-microplaten
Optically clear borosilicate glass bottom (light path 175±15 µm) and black PS frame.
  • All well formats available: 24, 96, 384 and 1536 wells
  • Standard SBS footprint
  • SensoPlate™ Plus option for high throughput screening applications
  • Free of DNase, RNase and human DNA, and non-pyrogenic
SensoPlate™ glass bottom microplates are designed for fluorescence correlation spectroscopy. The glass bottom allows transmission measurements in the wavelength range above 350 nm. Microscopic applications, such as confocal microscopy, are another potential area of application. The 175 µm thick glass bottom is equivalent to the light path of standard cover slips. SensoPlate™ are available sterile with lid, but should be tested for their suitability for cell culture before application.
SensoPlate™ Plus glass bottom microplates are intended for optical applications in high throughput screening. They are characterised by an improved geometry, which enables interference-free measurements and also microscopy over the whole surface of the bottom of the microplate.