Microtiterplaten, wit, 96-wells, Sterilin®
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
634-0007EA 180 EUR
SLIN611F96WT
Microtiterplaten, wit, 96-wells, Sterilin®
PS, non-treated. Ideal for bioluminescence studies.
  • White pigment aids reflectivity and sensitivity and prevents well-to-well cross talk
  • Chimney well design to help reduce contamination
Kleur: wit