Microtiterplaten, 96-wells, Sterilin®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sterilin®
634-0009EA 74.9 EUR
634-0009 634-0008 734-0482 734-0481 634-0010 734-0483
Microtiterplaten, 96-wells, Sterilin®
Microplaten HTS-microplaten
PS, non treated. Used for serology and microbiology. Compatible with automated plate readers and washers.
  • Suitable for use at a wavelength of 340 nm
  • Condensation rings on the lid (supplied separately) together with raised well rims on the base help minimise the risk of contamination from surrounding wells
  • Frosted write-on area on end wall, orientation corners and alpha numeric labelling ensure easy sample identification
  • Hanging well design ensures even temperature distribution around each well
  • Manufactured from virgin polystyrene for excellent optical clarity
  • Aseptically manufactured without surface treatment for a medium binding surface
  • Irradiated for sterility and a more hydrophilic surface, providing high binding
Kleur: helder
Certificaten: CE marked in accordance with the European Directive 98/72/EC.