Microarray plates, HTA™ Plate
Leverancier: Greiner bio-one
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HTA™
731-3019EA 1360 EUR
731-3019 731-3018 731-3020
Microarray plates, HTA™ Plate
Microplaten Microarray-platen
Plastic plates for high-throughput microarrays. With 96 shallow wells and a low well rim, the HTA™ Plate has been optimised for the simultaneous analysis of multiple samples.
  • Available with multifunctional, 3-D Amino, 3-D Aldehyde or 3-D Epoxy surface functionality
  • Partition of the plate into four individual strips containing 24 wells each gives flexibility, permitting processing of a variable number of samples each day
  • Plate dimensions are compatible with automated systems
  • Low well rim of only 0,3 mm
  • Low autofluorescence