Netherlands
Order Entry
Cover glasses, Nunc™ Lab-Tek™ II
Cover glasses, Nunc™ Lab-Tek™ II
   Sustainable Options Available
Certificates
undefined
Cover glasses, Nunc™ Lab-Tek™ II
   Sustainable Options Available
Specifications
  • Catalog No:
  • 734-2061
  • 734-2059
  • 734-2057
  • 734-2055
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Information
  • Catalog #
    Supplier #
    Volume
    Material
    Pk
    Availability
    Price
  • Cover glasses with one chamber (8,6 cm²)
    Cover glasses with one chamber (8,6 cm²)
    734-2055
    155360
    2 - 4,5 ml
    PS
    16
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 364,17 €
    364,17 €
    1 * 16 ST
       364,17 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Cover glasses with two chambers (4 cm²)
    Cover glasses with two chambers (4 cm²)
    734-2057
    155379
    1 - 2 ml
    PS
    16
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 366,67 €
    366,67 €
    1 * 16 ST
       366,67 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Cover glasses with four chambers (1,7 cm²)
    Cover glasses with four chambers (1,7 cm²)
    734-2059
    155382
    0,5 - 1 ml
    PS
    16
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 367,50 €
    367,50 €
    1 * 16 ST
       367,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Cover glasses with eight chambers (0,7 cm²)
    Cover glasses with eight chambers (0,7 cm²)
    734-2061
    155409
    0,2 - 0,5 ml
    PS
    16
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 375,00 €
    375,00 €
    1 * 16 ST
       375,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact