U heeft gezocht op: Mailer enveloppen en transportverpakkingen
23  results were found
SearchResultCount:"23"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Sonoco Thermosafe
Omschrijving: Polyurethane.
Leverancier: Sonoco Thermosafe
Omschrijving: EPS (expanded polystyrene).
Catalogus nummer: (SIMPM477-6)
Leverancier: Simport Scientific
Omschrijving: PVC, translucent.
UOM: 1 * 50 ST
Leverancier: Sonoco Thermosafe
Omschrijving: These moulded containers are designed for logistics applications in the pharmaceutical industries, where temperature-sensitive products have to be tra...
Sale
Leverancier: Sonoco Thermosafe
Omschrijving: FreezeTherm® dry ice shipping systems provide a readily available, convenient, and affordable method of protecting frozen products in transit. A wide ...
Catalogus nummer: (216-1294)
Leverancier: Sonoco Thermosafe
Omschrijving: EPS (expanded polystyrene) and Neopor.
UOM: 1 * 55 ST
Catalogus nummer: (SCATE280C)
Leverancier: Sonoco Thermosafe
Omschrijving: Nomadic® pre-qualified shippers deliver precise temperature protection for refrigerated and frozen pharmaceuticals, biological specimens, and samples....
UOM: 1 * 1 ST
Catalogus nummer: (216-1323)
Leverancier: Sonoco Thermosafe
Omschrijving: EPS (expanded polystyrene) en Neopor.
UOM: 1 * 19 ST
Catalogus nummer: (216-1337)
Leverancier: Sonoco Thermosafe
Omschrijving: EPS (geëxpandeerd polystyreen).
UOM: 1 * 4 ST
Catalogus nummer: (216-1339)
Leverancier: Sonoco Thermosafe
Omschrijving: EPS (uitgebreid polystyreen) en Neopor.
UOM: 1 * 35 ST
Catalogus nummer: (DGINBM003)
Leverancier: DGP INTELSIUS GMBH
Omschrijving: The Intelsius® Biomailer is designed for use within national and global postal networks shipping small specimens, such as blood vials and slides.
UOM: 1 * 50 ST
New Product
Catalogus nummer: (DAKL520300)
Leverancier: DAKLAPACK
Omschrijving: The patented MiniMailBox system is approved for use in every country as a recognised safe way of transporting biological substances category B, UN3373...
UOM: 1 * 100 ST
Leverancier: DAKLAPACK
Omschrijving: Blood transport blister, one position, PP, wit
Catalogus nummer: (DAKL20121)
Leverancier: DAKLAPACK
Omschrijving: Mini mailing boxes are designed for sending diagnostic samples of Category B.
UOM: 1 * 100 ST
Catalogus nummer: (DAKL460583)
Leverancier: DAKLAPACK
Omschrijving: These non-transparent absorbing materials are part of the packaging instruction P650. In emergencies the material will absorb all the liquid that has ...
UOM: 1 * 100 ST
Leverancier: DAKLAPACK
Omschrijving: These kits contain a cardboard outer box (UN3373 or UN2814), a 500 ml green leak proof container, plastic shock absorbers and absorbing tube holders.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 23
no targeter for Bottom