U heeft gezocht op: Schermen
11  results were found
SearchResultCount:"11"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (113-8820)
Leverancier: VWR Collection
Omschrijving: Acrylic shield, transparent, thickness: 10 mm, for protection from low energy beta radiation. The dual angled version is angled at both the bottom and...
UOM: 1 * 1 ST
Leverancier: Bel-Art Products, a Part of SP
Omschrijving: The stable transparent safety shield PC (thickness: 4,7 mm) is constructed of clear PC barrier and heavy steel base is coated with blue epoxy coating.
Catalogus nummer: (113-2500)
Leverancier: Bel-Art Products, a Part of SP
Omschrijving: These splash shields are designed to provide safety, comfort, flexibility and quality in a design that will adapt to your specific needs.
UOM: 1 * 1 ST
Catalogus nummer: (113-2801)
Leverancier: Thermo Fisher Scientific
Omschrijving: PC/acetaat met magnetische basis, transparant, dikte: 3 mm. Veiligheidsspatscherm met flexibele arm bevestigt zich eenvoudg aan tafels, rekken, m...
UOM: 1 * 1 ST
Catalogus nummer: (KART571)
Leverancier: KARTELL
Omschrijving: PMMA protection shield, transparent, thickness: 5 mm, provides laboratory and healthcare personnel with facial protection from hazardous substances, a...
UOM: 1 * 1 ST
Leverancier: Thermo Fisher Scientific
Omschrijving: Acryl, transparant, dikte: 9 mm
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Omschrijving: Protective shield made from acrylic glass to be used as splinter shield and splash guard.
Catalogus nummer: (BELAF249760003)
Leverancier: Bel-Art Products, a Part of SP
Omschrijving: Acrylic protection shield; transparent, thickness 9,5 mm
UOM: 1 * 1 ST
Leverancier: Thermo Fisher Scientific
Omschrijving: Acryl, transparant, dikte: 9 mm.
Catalogus nummer: (KART572)
Leverancier: KARTELL
Omschrijving: Acryl beschermkap, transparant, dikte: 9 mm
UOM: 1 * 1 ST
Leverancier: Roth Carl
Omschrijving: These safety shields are made from transparent acrylic glass of 8 mm thick.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 11 of 11
no targeter for Bottom