Small stand with circular base (D)
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
241-0078EA 50.4 EUR
VWRF08465.087
Small stand with circular base (D)
Cast iron stand, Ø 90 mm, with rod (8×250 mm).
  • With central hole for rod
  • Robust and stable
  • Good chemical resistance