Small stand with H-type base (C)
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
241-0115EA 46.3 EUR
VWRF08545.084
Small stand with H-type base (C)
Stand with H-formed base and steel rod (8×250 mm). The base (125×50 mm) is made of nickel-plated and laquered nodular cast iron.
  • Central hole for rod
  • Robust and stable
  • Good chemical resistance