U heeft gezocht op: Scalpels
74  results were found
SearchResultCount:"74"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: SWANN-MORTON
Omschrijving: Met roestvrijstalen mesje en kunststof handvat.
Leverancier: SWANN-MORTON
Omschrijving: Koolstofstaal.
Leverancier: SWANN-MORTON
Omschrijving: These stainless steel blades are thicker than a standard blade thus offering additional rigidity and strength when used in conjunction with the unique...
Leverancier: SWANN-MORTON
Omschrijving: Scalpel blades, Roestvrij staal, Steriel, SM 62
Leverancier: SWANN-MORTON
Omschrijving: The ACM tool set is presented in a resilient blue carry case and contains a comprehensive range of 13 precision ground carbon steel blades manufacture...
Leverancier: SWANN-MORTON
Omschrijving: Stainless steel. The 158 mm skin graft blades are used in the surgical removal of a patch of healthy skin from one area of the patient's body, usually...
Leverancier: SWANN-MORTON
Omschrijving: Scalpel blades, Koolstofstaal, Steriel, No. 25A, Individual foil packs, carbon steel
Leverancier: SWANN-MORTON
Omschrijving: Roestvrij staal.
Leverancier: SWANN-MORTON
Omschrijving: Roestvrij staal.
Leverancier: SWANN-MORTON
Omschrijving: These non sterile disposable scalpels consist of a stainless steel blade and a PS handle.
Catalogus nummer: (SWAN6061)
Leverancier: SWANN-MORTON
Omschrijving: Stainless steel.
UOM: 1 * 1 ST
Leverancier: SWANN-MORTON
Omschrijving: Surgical quality blades for use with handles No. 5A and No. 6A.
Leverancier: SWANN-MORTON
Omschrijving: Stainless steel blade, plastic handle. These disposable safety scalpels support and enhance safe passing procedures and protocols already practiced wi...
Leverancier: SWANN-MORTON
Omschrijving: These acrylic craft knife handles have a stainless blade fitting.
Leverancier: SWANN-MORTON
Omschrijving: Stainless steel blade, sterile.
Leverancier: SWANN-MORTON
Omschrijving: These handles and blades are ideal for artwork, craft work and modelling.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 74
no targeter for Bottom