VWR®, Mini Equipment Log Books
Supplier: VWR Collection
Certificates
About this item
'FLEX' covers with coating, for durability in laboratory environments. Acid-free, archive-safe paper (90 gsm).
- Pages specifically designed for logging and recording equipment data
- Tough coated cover
Format: A5 (14,8×21 cm) or A6 (10,5×14,8 cm), 96 numbered pages.
All books made in accordance with GLP, GMP and GCP guidelines.
Certifications: All books made in accordance with GLP, GMP and GCP guidelines.
Related Information
Page Samples VWR Log Book 2016.pdf VWR Laboratory Notebooks VWR® for Calibrations VWR® for Weighing VWR® For Your Space
Product Literature
More
TDS_VWR_MINI_EQUIPMENT_LOG_BOOKS.pdf
Product specification
Product Information
- Catalog #ColourPaper sizeNo. of pagesPkAvailabilityPrice
- Mini equipment log bookLocal Article Number: VWRI818-0108Specifications:
Specifications
Environmentally Preferable - Mini equipment log bookLocal Article Number: VWRI818-0109Specifications:
Specifications
Environmentally Preferable
Frequently Bought Together