U heeft gezocht op: Sterilisatie-indicatoren
57  results were found
SearchResultCount:"57"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Merck
Omschrijving: Sterikon® plus bio-indicatoren kunnen worden gebruikt om de functionaliteit van autoclaven te testen (15 minuten bij 121 °C). Ze kunnen ook worden geb...
Leverancier: LIOFILCHEM S.R.L.
Omschrijving: Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Leverancier: LIOFILCHEM S.R.L.
Omschrijving: Biological indicators for validating the sterilisation using steam (autoclaves), VHP (isolators), dry heat and irradiation processes.
Leverancier: LIOFILCHEM S.R.L.
Omschrijving: Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Leverancier: LIOFILCHEM S.R.L.
Omschrijving: Biological indicators for validating the sterilisation using VHP in isolators.
Leverancier: MICRONOVA
Omschrijving: Saturated crepe paper/synthetic poly isoprene adhesive in plastic core. It is a proven method of positively confirming a completed sterilisation cycle...
Veiligere gezondheid van mens en milieu
Leverancier: LIOFILCHEM S.R.L.
Omschrijving: Biological indicators for validating sterilisation using steam sterilisation (autoclaves).
Leverancier: 3M MEDICAL
Omschrijving: An exposure monitor that securely seals packs, and provides a visual assurance that the package has been exposed to the steam sterilisation process.
Catalogus nummer: (804-0226)
Leverancier: 3M MEDICAL
Omschrijving: This chemical indicator tape is designed for use as packaging tape and for the immediate identification of items exposed to dry heat processing.
UOM: 1 * 1 ROL
Catalogus nummer: (BRND61750)
Leverancier: Brand
Omschrijving: This self-adhesive crepe indicator tape turns from white to brown upon autoclaving.
UOM: 1 * 1 ST
Catalogus nummer: (481-0842)
Leverancier: 3M MEDICAL
Omschrijving: Integrating indicator for use in determining whether the process conditions necessary for sterilisation were met inside each pack. Can be used in all ...
UOM: 1 * 500 ST
Catalogus nummer: (489-0109)
Leverancier: 3M MEDICAL
Omschrijving: Indicator tapes are Class 1 process indicators used for exposure monitoring. They provide assurance to the operator handling the processed items that ...
UOM: 1 * 1 ROL
Catalogus nummer: (481-0844)
Leverancier: 3M MEDICAL
Omschrijving: This steam chemical indicator offers a distinct off-white to dark brown/black color change when exposed to steam sterilization.
UOM: 1 * 240 ST
Catalogus nummer: (817-0302)
Leverancier: Texwipe
Omschrijving: Sterilisatie-indicatoren, TX522A, Cleanroom labels, autoclaveerbaar, Lengte: 50 mm, Breedte: 50 mm
UOM: 1 * 1 ROL
Catalogus nummer: (489-1310)
Leverancier: KARTELL
Omschrijving: Plakband.
UOM: 1 * 1 ROL
Leverancier: PRECISION DYNAMICS CORP.-TIMEM
Omschrijving: Witte kleefband van crêpepapier, voor gebruik in standaardautoclaven.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 57
no targeter for Bottom