Spatbollen, Stutzer, recht
Leverancier: Lenz
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
271-2233EA 60.3 EUR
271-2233 201-2232 201-1066 201-0342
Spatbollen, Stutzer, recht
Onderdelen voor destillatie Destillatiekoppen
DURAN®, borosilicate 3.3 glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Place directly onto the distillation flask to intercept droplets, with two standard ground joints.