U heeft gezocht op: Reservoirs
21  results were found
SearchResultCount:"21"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: BÃœRKLE
Omschrijving: These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
Leverancier: BÃœRKLE
Omschrijving: Containers in LDPE, transparent, with lids, for various applications. Produced seamlessly and stress relieved with the LaboPlast® sintering method.
Catalogus nummer: (ENTEE14-35-01-0215)
Leverancier: ENTEGRIS INC
Omschrijving: Injection molded round tank, in PFA material.
UOM: 1 * 1 ST
Catalogus nummer: (ENTEE92-01D-0215)
Leverancier: ENTEGRIS INC
Omschrijving: Injection molded rectangular tanks with dual capacity.
UOM: 1 * 1 ST
Leverancier: Thermo Fisher Scientific
Omschrijving: Voordelige tanks met schaalverdeling. Met uitstekende rand (flens). Wanddikte ~2,4 mm. LLDPE, doorschijnend met deksel en kraan.
Sale
Leverancier: Thermo Fisher Scientific
Omschrijving: Doorzichtig LLDPE, met deksel. Voordelige tanks met schaalverdeling. Met uitstekende rand (flens). Wanddikte ∼2,4 mm. Tank 216-0322 kan ook worden ge...
Leverancier: Thermo Fisher Scientific
Omschrijving: Heavy-duty cylindrical LLDPE tanks are offered in sizes ranging from 19 to 757 liters and include covers to signficantly reduce evaporation and contam...
Leverancier: Thermo Fisher Scientific
Omschrijving: These heavy-duty rectangular LLDPE tanks with cover can withstand most acid, alcohols and bases. Tanks feature stepped flange at top provides reinforc...
Leverancier: Thermo Fisher Scientific
Omschrijving: These PP rectangular tanks with cover can significantly reduce evaporation and contamination.
Leverancier: Thermo Fisher Scientific
Omschrijving: Cilindrische tank, Materiaal: LDPE, Inhoud: 208 l, Ø uitw.×h: 560×914 mm
Leverancier: Thermo Fisher Scientific
Omschrijving: Cylindrical tank, Materiaal: PP, Inhoud: 19 l, Ø uitw.×h: 280×380 mm
Sale
Leverancier: Thermo Fisher Scientific
Omschrijving: These dome tanks are designed for use as closed containment systems. Tanks feature a 150 mm neck opening fitted with a gasketed closure for secure sea...
Leverancier: GLASWARENFABRIK KARL HECHT
Omschrijving: Tanks van natronkalkglas voor multifunctioneel gebruik.
Leverancier: Bel-Art Products, a Part of SP
Omschrijving: These seamless, rotational-molded PE tanks are lightweight and easy-to-handle.
Leverancier: MARIENFELD
Omschrijving: Aquariumtanks voor een groot aantal toepassingen. Gemaakt van helder sodaglas.
Leverancier: ELGA LabWater
Omschrijving: Pure and ultra pure water system, Opslagreservoir, 30 l, 376 × 340 × 660 mm
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 21
no targeter for Bottom