Alcoholteststrips voor Salvia
Leverancier: SureScreen Diagnostics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-1245EA 143 EUR
720-1245
Alcoholteststrips voor Salvia
Drugstests
For single use, occasional or at incident alcohol testing. The saliva alcohol test strip is a rapid, highly sensitive method to detect the presence of alcohol in saliva and provide an approximation of relative blood alcohol concentration (BAC) at 0,02% or greater. Detection ranges from 0,02% through to 0,30% for the approximate relative blood alcohol concentration. Ideal for one-off occasions and perfect for off-site checks.
  • Accurate and quantitative result with saliva
  • No false positives
  • Results in two minutes
  • Can be used by non medical staff