U heeft gezocht op: Dialysebuisjes
8  results were found
SearchResultCount:"8"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Thermo Fisher Scientific
Omschrijving: Single-Use RED (rapid equilibrium dialysis) Plate is composed of disposable high-density polypropylene and is pre-loaded with 48 equilibrium dialysis ...
Leverancier: Thermo Fisher Scientific
Omschrijving: Pierce™ low protein binding microcentrifuge tubes help to minimise protein sample loss during sample manipulation and are ideal in applications where ...
Leverancier: G-Biosciences
Omschrijving: Tube‐O‐Reactor™, Micro, for 20 to 250 µl samples or Medi for 0,2 to 2 ml samples.
Leverancier: Cytiva
Omschrijving: The disposable tubes in mini dialysis kit offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit eac...
Leverancier: Merck Millipore (Novagen)
Omschrijving: D-Tube96™ Dialysers allow convenient, high throughput dialysis of 96 samples simultaneously. The device features the advantages of the D-Tube™ Dialyze...
Leverancier: G-Biosciences
Omschrijving: 'NO-LOSS' dialyser.
Catalogus nummer: (786-142-45MC)
Leverancier: G-Biosciences
Omschrijving: Genomic Tube‐O‐DIALYZER™ (0.45µm). 
UOM: 1 * 1 Kit
Leverancier: G-Biosciences
Omschrijving: Tube-O-CONCENTRATOR™ Mini for 20 to 250 μl volumes or Medi for 0,2 to 2,5 ml volumes.
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8 of 8
no targeter for Bottom