Slangen, neopreen, Norprene® A-60-F
Leverancier: Saint-Gobain
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VERNR6F12-43EA 280 EUR
VERNR6F12-43
Slangen, neopreen, Norprene® A-60-F
Formulation A-60-F, cream.
  • Long flex life in peristaltic pumps
  • Compatible with virtually all common sanitizers and cleaners
  • Ozone and UV light resistant
  • Repeatedly autoclavable
  • Chemically compatible with a wide range of fluids
Food process tubing is ideal for use in clean-in-place and steam-in-place cleaning and sterilisation systems.
Temperature resistance: −60 to +135 °C
Hardness (Shore durometer A): 61
Density: 0,98
Certificaten: Complies FDA 21 CFR, 177.2600 3-A Sanitary Standards and NSF Standard 51.