Glasbuizen
Leverancier: Schott AG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCOR1193950EA 325 EUR
SCOR1193950 SCOR1193957 SCOR1193956 SCOR1193955 SCOR1193954 SCOR1193953 SCOR1193952 SCOR1193951 SCOR1193959 SCOR1027917 SCOR1193961 SCOR1193960 SCOR1193606 SCOR1216929 SCOR1545306 SCOR1193609 SCOR301.9AR SCOR1193608 SCOR1193607 SCOR1193614 SCOR1193613 SCOR1193612 SCOR1028436 SCOR1193611 SCOR1193610 SCOR1028427 SCOR1579029 SCOR1545456 SCOR1193626 SCOR1193625 SCOR1193469 SCOR1193468 SCOR1193467 SCOR1027602 SCOR1193909 SCOR1193908 SCOR1548718 SCOR1193474 SCOR1485593 SCOR1193473 SCOR1193472 SCOR1552679 SCOR1193914 SCOR1028254 SCOR1193518 SCOR1193517 SCOR1193913 SCOR1116951 SCOR1193912 SCOR1193515 SCOR1193911 SCOR1309479 SCOR1193910 SCOR1588626 SCOR1028407 SCOR1193519 SCOR1193488 SCOR1193521 SCOR1193487 SCOR1193520 SCOR1193524 SCOR1193523 SCOR1193522 SCOR1193247 SCOR1193491 SCOR1185432 SCOR1553423 SCOR1193252 SCOR1193494 SCOR1193251 SCOR1193493 SCOR1193492 SCOR1193936 SCOR1193538 SCOR1193537 SCOR1193535 SCOR1028631 SCOR1027932 SCOR1027934 SCOR1027936 SCOR1193939 SCOR1027935 SCOR1027938 SCOR1193938 SCOR1193937 SCOR1193540 SCOR1193947 SCOR1193946 SCOR1193945 SCOR1193944 SCOR1193943 SCOR1027930 SCOR1028347 SCOR1193949 SCOR1193948 SCOR1027929 SCOR1027928 SCOR1404404
Glasbuizen
Slangen Transfer Tubing
AR-Glas®, clear.
  • Good chemical and physical properties
  • Suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals
  • Not resistant to high thermal loads
Tube length ~1500 mm.