Ubbelohde viscometers with constant
Supplier: SI Analytics
Certificates
About this item
Calibrated for automatic measurement, viscometers with suspended ball level for determination of absolute and relative viscosity of liquids with Newtonian flow behaviour.
Recommended minimum flow through time is 200 seconds.
Certifications: DIN 51562-1, ISO/DIS 3105 (BS-IP-SL).
Specifications
- Catalog No:
- 616-3341
- 616-3340
- 616-3336
- 616-3345
- 616-3335
- 616-3344
- 616-3338
- 616-3343
- 616-3339
- 616-3342
- Length:
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- Volume:
- 15 − 75 ml
- 15 − 75 ml
- 15 − 75 ml
- 15 − 75 ml
- 15 − 75 ml
- 15 − 75 ml
- 15 − 75 ml
- 15 − 75 ml
- 15 − 75 ml
- 15 − 75 ml
Specifications
Product Information
- Catalog #Supplier #RangeConstantLengthØ int.VolumePkAvailabilityPrice
- 616-33382854002010,5 - 3 mm²/s0,003 K290 mm0,47 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400201Local Article Number: SCOI285400201Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml - 616-33392854002180,8 - 5 mm²/s0,005 K290 mm0,53 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400218Local Article Number: SCOI285400218Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml - 616-33402854002261,2 - 10 mm²/s0,01 K290 mm0,63 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400226Local Article Number: SCOI285400226Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml - 616-33412854002343 - 30 mm²/s0,03 K290 mm0,84 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400234Local Article Number: SCOI285400234Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml - 616-33352854001725 - 50 mm²/s0,05 K290 mm0,95 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400172Local Article Number: SCOI285400172Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml - 616-334228540024210 - 100 mm²/s0,1 K290 mm1,13 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400242Local Article Number: SCOI285400242Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml - 616-334328540025930 - 300 mm²/s0,3 K290 mm1,50 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400259Local Article Number: SCOI285400259Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml - 616-333628540018950 - 500 mm²/s0,5 K290 mm1,69 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400189Local Article Number: SCOI285400189Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml - 616-3344285400267100 - 1000 mm²/s1 K290 mm2,01 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400267Local Article Number: SCOI285400267Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml - 616-3345285400275300 - 3000 mm²/s3 K290 mm2,65 mm15 − 75 ml1Log in for availability1 * 1 ST483,00 €Supplier Article Number: 285400275Local Article Number: SCOI285400275Specifications:
More
Specifications
Length290 mmEnvironmentally PreferableVolume15 − 75 ml