Dri-block® thermostaten, serie DB
Leverancier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Techne® Dri-block®
460-0058EA 1137.22 EUR
460-0058 460-0057 460-0054
Dri-block® thermostaten, serie DB
Blokverwarmers Analoge blokverwarmers
The Techne® Dri-Blocks® are particularly suitable for microbiology and clinical laboratories for incubation, boiling, inactivation, wet ashing, sample concentration and enzyme analysis. There is a choice of temperature ranges; 25 to 100 °C or 25 to 200 °C. These models have a rotary knob for temperature setting (but no digital display).
  • Wide range of interchangeable blocks to accommodate tubes, microcentrifuge tubes and 96-well plates
  • Very accurate temperature control
  • Temperature stability at 40 °C is ±0,05 or ±0,1 °C, maximum temperature variation between identical blocks is ±0,2 °C
  • Rapid heat up rate: 8 minutes from 30 to 37 °C
Certificaten: Beschermingsklasse conform DIN EN 60529: IP 41
Opgelet: Note: Temp. stability value shown above is at 100 °C.
Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar technical.service.be@vwr.com