Microhematocrietlezer
Leverancier: Hawksley
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-0100EA 0 EUR
720-0100
Microhematocrietlezer
Diagnostische systemen Clinical Analysers
For determination of packed cell volume, using 75 mm capillary tubes with total fluid column heights between 40 and 70 mm.