Elektronische formuletellers, DIFF 15
Leverancier: CAT (M. Zipperer GmbH)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-0814EA 1770 EUR
CATA60244
Elektronische formuletellers, DIFF 15
  • Maximum telling tot 1000
  • Aflezing in % of van de reële waarde
  • Ingebouwde herlaadbare batterij
Four programmes: LEUKO, MYELO/RBC, RETI, FREE Choice.