Netherlands
Order Entry
Digital burettes, Top Buret
Digital burettes, Top Buret
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
undefined
Digital burettes, Top Buret
Supplier:  EPPENDORF
   Sustainable Options Available
Specifications
  • Catalog No:
  • 613-3667
  • 613-3668
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Information
  • Catalog #
    Supplier #
    Capacity
    Division
    Accuracy
    Imprecision
    Pk
    Availability
    Price
  • Top Buret M
    Top Buret M
    613-3667
    4965000.017
    25 ml
    0,01 ml
    ±0,2 %
    <0,1 %
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 990,00 €
    990,00 €
    1 * 1 ST
       990,00 €
    Supplier Article Number: 4965000.017
    Local Article Number: EPPE4965000.017
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Top Buret H
    Top Buret H
    613-3668
    4965000.025
    50 ml
    0,01 ml
    ±0,2 %
    <0,1 %
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 990,00 €
    990,00 €
    1 * 1 ST
       990,00 €
    Supplier Article Number: 4965000.025
    Local Article Number: EPPE4965000.025
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
    Environmental certification