Conductivity cells for conductivity meters, Jenway®
Leverancier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
663-0247EA 573 EUR
663-0247 663-0005 665-0003 JENW027113 665-0004
Conductivity cells for conductivity meters, Jenway®
Elektroden Geleidbaarheidselektroden
Conductivity cells with built-in ATCs for use with Jenway portable conductivity meter (model 470) and/or Jenway bench conductivity meter (model 4510 and 4520).
  • Keep measurements accurate with replacement probes
  • Probes are 120 mm in length, have a 12 mm diameter, and 1 m long cable
If your conductivity measurements are off, you might need a new cell, sensor, or calibration kit. Update, upgrade, or replace to get the best out of your Jenway® conductivity meter.