Netherlands
Order Entry
Combs for dual-gel electrophoresis systems, Owl™ P9DS, P8DS and P10DS
Combs for dual-gel electrophoresis systems, Owl™ P9DS, P8DS and P10DS
Certificates
undefined
Combs for dual-gel electrophoresis systems, Owl™ P9DS, P8DS and P10DS
Standard combs for P9DS
Product Information
  • Catalog #
    Supplier #
    Type
    Well volume
    Pk
    Availability
    Price
  • Standard comb, 10×0,8 mm teeth
    Standard comb, 10×0,8 mm teeth
    730-0266
    P1-10C
    Standard
    160 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 96,80 €
    96,80 €
    1 * 1 ST
       96,80 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 10×1,5 mm teeth
    Standard comb, 10×1,5 mm teeth
    730-0267
    P1-10D
    Standard
    305 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €
    Specifications:
    Specifications
    Environmentally Preferable
Standard combs for P8DS
Product Information
  • Catalog #
    Supplier #
    Type
    Well volume
    Pk
    Availability
    Price
  • Standard comb, 12×0,5 mm teeth
    Standard comb, 12×0,5 mm teeth
    730-0248
    MP-12A
    Standard
    30 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 12×0,8 mm teeth
    Standard comb, 12×0,8 mm teeth
    730-0249
    MP-12C
    Standard
    50 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 79,30 €
    79,30 €
    1 * 1 ST
       79,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 12×1,5 mm teeth
    Standard comb, 12×1,5 mm teeth
    730-0250
    MP-12D
    Standard
    95 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 8×1,5 mm teeth
    Standard comb, 8×1,5 mm teeth
    730-0258
    MP-8D
    Standard
    170 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 20×0,5 mm teeth
    Standard comb, 20×0,5 mm teeth
    730-0252
    MP-20A
    Standard
    20 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 15×0,5 mm teeth
    Standard comb, 15×0,5 mm teeth
    730-0251
    MP-15A
    Standard
    30 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 10×0,5 mm teeth
    Standard comb, 10×0,5 mm teeth
    730-0245
    MP-10A
    Standard
    40 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 8×0,5 mm teeth
    Standard comb, 8×0,5 mm teeth
    730-0256
    MP-8A
    Standard
    55 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 10×0,8 mm teeth
    Standard comb, 10×0,8 mm teeth
    730-0246
    MP-10C
    Standard
    65 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 79,30 €
    79,30 €
    1 * 1 ST
       79,30 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 6×0,5 mm teeth
    Standard comb, 6×0,5 mm teeth
    730-0253
    MP-6A
    Standard
    80 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 8×0,8 mm teeth
    Standard comb, 8×0,8 mm teeth
    730-0257
    MP-8C
    Standard
    90 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 10×1,5 mm teeth
    Standard comb, 10×1,5 mm teeth
    730-0247
    MP-10D
    Standard
    125 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 6×0,8 mm teeth
    Standard comb, 6×0,8 mm teeth
    730-0254
    MP-6C
    Standard
    130 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 6×1,5 mm teeth
    Standard comb, 6×1,5 mm teeth
    730-0255
    MP-6D
    Standard
    245 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,20 €
    82,20 €
    1 * 1 ST
       82,20 €
    Specifications:
    Specifications
    Environmentally Preferable
Standard combs for P10DS
Product Information
  • Catalog #
    Supplier #
    Type
    Well volume
    Pk
    Availability
    Price
  • Standard comb, 15×1,5 mm teeth
    Standard comb, 15×1,5 mm teeth
    730-0286
    P2-15D
    Standard
    245 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 25×0,8 mm teeth
    Standard comb, 25×0,8 mm teeth
    730-0294
    P2-25C
    Standard
    60 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 20×0,8 mm teeth
    Standard comb, 20×0,8 mm teeth
    730-0288
    P2-20C
    Standard
    85 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 25×1,5 mm teeth
    Standard comb, 25×1,5 mm teeth
    730-0295
    P2-25D
    Standard
    115 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 15×0,8 mm teeth
    Standard comb, 15×0,8 mm teeth
    730-0285
    P2-15C
    Standard
    130 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 20×1,5 mm teeth
    Standard comb, 20×1,5 mm teeth
    730-0289
    P2-20D
    Standard
    165 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 10×0,8 mm teeth
    Standard comb, 10×0,8 mm teeth
    730-0283
    P2-10C
    Standard
    215 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard comb, 10×1,5 mm teeth
    Standard comb, 10×1,5 mm teeth
    730-0284
    P2-10D
    Standard
    405 µl
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €
    Specifications:
    Specifications
    Environmentally Preferable