U heeft gezocht op: Kasten met laminaire stroming
10  results were found
SearchResultCount:"10"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: LABCONCO
Omschrijving: Clean benches provide product protection from environmental contaminants for applications requiring a particulate-free work area, including plant tiss...
New Product
Leverancier: CleanAir by Baker
Omschrijving: Draagframe voor CLF 575, in hoogte verstelbaar
Leverancier: CleanAir by Baker
Omschrijving: CleanAir by Baker DLF series is a high quality vertical laminar flow workbench range for handling non-hazardous materials that require a clean working...
Leverancier: Azbil Telstar Technologies
Omschrijving: Aeolus V Serie is a high quality vertical laminar flow workbench range for handling non-hazardous materials that require a particle free environment. ...
Leverancier: Azbil Telstar Technologies
Omschrijving: Aeolus H Serie is a high quality horizontal laminar flow workbench range for handling non-hazardous materials that require a particle free environment...
Leverancier: Thermo Scientific
Omschrijving: Laminaire flowkast, Heraguardâ„¢ ECO, Reinigingsbank, Heraguardâ„¢ ECO 1,8, Laminaire stroming: Horizontaal, Luchtsnelheid: 0,15 - 0,25 m/s (level I): 0,36 - 0,45 m/s (level II), Verlichting: >1000 lux, Geluidsniveau: <55 dB(A) (niveau I): <60 dB(A) (niveau II), Inwendige b×d×h: 1820×585×645 mm, Gewicht: 150 kg
Leverancier: EUROCLONE S.P.A.
Omschrijving: AURA HZ is an horizontal laminar air flow cabinet Class 100, for product protection from external contamination and cross-contamination inside the wor...
Catalogus nummer: (EUCLLV30500)
Leverancier: EUROCLONE S.P.A.
Omschrijving: Aura Miniâ„¢ Cabinets are compact down-flow cabinets, provide an ultimate cleanliness Class 100 work area where the highest safety for the products is a...
UOM: 1 * 1 ST
Catalogus nummer: (TELR1923477)
Leverancier: Azbil Telstar Technologies
Omschrijving: Werkbanken met verticale laminaire doorstroming zijn een ideale oplossing voor het hanteren van onschadelijke stoffen en ongevaarlijke biologische mon...
UOM: 1 * 1 ST
New Product
Leverancier: LABCONCO
Omschrijving: These PuriCare Open Access Transfer Station offer utmost efficiency for cage changing operations. Open access on two sides provides ample workspace fo...
Duurzame materialen New Product
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 10 of 10
no targeter for Bottom