Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Toebehoren voor Clarus® 500 GC
Leverancier: PERKIN ELMER GC-INSTRUMENTS
Order
Expand All / Collapse All
Omschrijving VPE VWR catalogus nummer Hoeveelheid
Replacement column nut, 1/16”, connects capillary column to adapter 5 PERC09903392
Photoionization detector (PID), add on kit, 230 V, for Clarus® 500 / Clarus® 600 1 PERCN6520027
Sealing resin, for polyimide column connector 5 g PERCN9301343
Interface NCI 901 (acquisitie-interface analoog / digitale mono-track) 1 568-0622
IPM-sturing AS XL / Clarus® 500 / Clarus® 600 1 568-0528
TotalChrom-software voor multi-instrumentenomgeving 1 568-0527
Update voor TotalChrom 6x en TotalChrom 6.3 1 568-0529
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid
Een nieuwe uitstraling en een hernieuwde belofte
Gebouwd op de kenmerken & middelen die u kent, geoptimaliseerd met de innovatie van Avantor ®. Ons nieuwe merk symboliseert flexibiliteit en ons vermogen om ons aan de behoeften van de klant aan te passen.
KOM MEER TE WETEN BEGIN MET WINKELEN