Homogeniser disposable pestles and tubes, polypropene, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCERSP749540-0000EA 328 EUR
SCERSP749540-0000
Homogeniser disposable pestles and tubes, polypropene, KIMBLE®
Homogenisatoren Draagbare homogenisatoren
Stainless steel cordless drive unit. Pressure is 14.7 PSI.
  • Resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
  • Operates at 2000 to 3000 min⁻¹ with fresh batteries and no load
Leveringsinformatie: Supplied with 2× AA batteries.