Slotted lids for ADP polarimeters
Leverancier: Bellingham + Stanley
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
635-0416EA 232 EUR
635-0416 635-0417 635-0418 635-0419
Slotted lids for ADP polarimeters
Polarimeters
These slotted lids provide stability to funnel through tubes whilst ensuring no extraneous light enters the measurement chamber.
Suitable for flow through applications in flavours and fragrance industries.