U heeft gezocht op: Eiwitzuivering
296  results were found
SearchResultCount:"296"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: Merck
Omschrijving: Ion Exchanger I is a strong acid cation exchangers widely used in separation of noble earths and amino acids, water treatment, analysis and food indus...
Leverancier: Merck
Omschrijving: 50% aqueous suspension in 1 M NaCl.
Catalogus nummer: (SUPLG50150-50G)
Leverancier: Merck
Omschrijving: Sephadex® G-50 is a gel filtration medium used in affinity chromatography, protein chromatrography and gel filtration chromatography.
UOM: 1 * 50 g
Leverancier: Avantor
Omschrijving: Media op silicabasis die worden gebruikt voor de zuivering van eiwitten op basis van differentiële zoutgeïnduceerde hydrofobiciteit. De gemengde modus...
Leverancier: Avantor
Omschrijving: J.T.Baker® BAKERBOND®-ondersteunde platen voor vloeistofextractie (SLE) zijn een ideaal en sneller alternatief voor traditionele vloeistof-vloeistofex...
Leverancier: Avantor
Omschrijving: The J.T.Baker® BAKERBOND® PROchievA™ protein A resin offers superior dynamic binding capacity with excellent alkaline stability ...
Leverancier: Avantor
Omschrijving: Dankzij de J.T.Baker® BAKERBOND® eiwitneerslagplaten kunnen eiwitten snel uit biologische monsters worden verwijderd.
Catalogus nummer: (786-789)
Leverancier: G-Biosciences
Omschrijving: Immobilised Pepstatin is for the purification of cathepsins, pepsin, bacterial aspartic proteases, HIV proteases and other molecules that bind pepstat...
UOM: 1 * 5 mL
Leverancier: G-Biosciences
Omschrijving: For the rapid purification of soluble, 6X His or GST tagged proteins.
Leverancier: G-Biosciences
Omschrijving: Nickel chelating resin, Type: Liquid
Leverancier: Rockland Immunochemicals
Omschrijving: Protein G alkaline phosphatase (calf intestine)
Leverancier: AGILENT
Omschrijving: The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as...
Leverancier: G-Biosciences
Omschrijving: Immobilised Protein A and G are designed for binding the constant domains of immunoglobulin (Ig) molecules. Immobilised Protein A is coupled to 6% hig...
Leverancier: G-Biosciences
Omschrijving: Sulphhydryl coupling resin, 5×2 ml columns
Catalogus nummer: (786-803)
Leverancier: G-Biosciences
Omschrijving: Pearlâ„¢ Antibody Clean Up Kit, for removal of BSA and gelatin, Pearlâ„¢
UOM: 1 * 1 Kit
Catalogus nummer: (IMMRIR1015)
Leverancier: ImmunoReagents
Omschrijving: ImmunoReagents Sheep IgG Affinity Prep resins are prepared using highly purified species specific immunoglobulins. This resin is suitable for removal ...
UOM: 1 * 2 mL
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 16 of 296
no targeter for Bottom